Assay Design



Gene Transcripts

Gene Symbol
BCHE
Entrez Gene ID
534616
Gene Name
butyrylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.1: 101522670 - 101601155 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076906.2 NP_001070374.1 1 - 2 105 106
GenBank mRNA BC123600.1 - 1 - 2 107 106

View Full Product Details