Assay Design



Gene Transcripts

Gene Symbol
NMS
Entrez Gene ID
768331
Gene Name
neuromedin S
Gene Aliases
-
Chromosome Location
Chr.11: 5537457 - 5548804 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077522.2 NP_001070990.1 6 - 7 424 89
GenBank mRNA BC120470.1 - 6 - 7 425 89

View Full Product Details