Assay Design



Gene Transcripts

Gene Symbol
CBX8
Entrez Gene ID
506458
Gene Name
chromobox 8
Gene Aliases
-
Chromosome Location
Chr.19: 52742564 - 52745101 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077862.1 NP_001071330.1 4 - 5 255 66
GenBank mRNA AY962104.1 - 3 - 4 168 66
BC120326.1 - 4 - 5 255 66

View Full Product Details