Assay Design



Gene Transcripts

Gene Symbol
LHFPL6
Entrez Gene ID
532944
Gene Name
LHFPL tetraspan subfamily member 6
Gene Aliases
LHFP
Chromosome Location
Chr.12: 22796913 - 23031263 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077990.1 NP_001071458.1 2 - 3 897 53
GenBank mRNA BC120189.1 - 2 - 3 897 53

View Full Product Details