Assay Design



Gene Transcripts

Gene Symbol
FAIM
Entrez Gene ID
616795
Gene Name
Fas apoptotic inhibitory molecule
Gene Aliases
-
Chromosome Location
Chr.1: 130488923 - 130505390 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079626.2 NP_001073094.1 4 - 5 552 101
GenBank mRNA BC122668.1 - 4 - 5 553 101

View Full Product Details