Assay Design



Gene Transcripts

Gene Symbol
KCNJ12
Entrez Gene ID
538479
Gene Name
potassium inwardly rectifying channel subfamily J member 12
Gene Aliases
IRK2, KIR2.2
Chromosome Location
Chr.19: 35328131 - 35365898 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024690.2 NP_001019861.2 1 - 2 70 104
GenBank mRNA BC133380.1 - 1 - 2 70 104

View Full Product Details