Assay Design



Gene Transcripts

Gene Symbol
CENPC
Entrez Gene ID
450214
Gene Name
centromere protein C
Gene Aliases
CENPC1
Chromosome Location
Chr.6: 83183412 - 83277556 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045881.2 NP_001039346.2 13 - 14 2403 101
GenBank mRNA AY693782.1 - 4 - 5 368 101
BC105482.1 - 13 - 14 2405 101

View Full Product Details