Assay Design



Gene Transcripts

Gene Symbol
CENPC
Entrez Gene ID
450214
Gene Name
centromere protein C
Gene Aliases
CENPC1
Chromosome Location
Chr.6: 83183412 - 83277556 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045881.2 NP_001039346.2 15 - 16 2633 121
GenBank mRNA AY693782.1 - 6 - 7 598 121
BC105482.1 - 15 - 16 2635 121

View Full Product Details