Assay Design



Gene Transcripts

Gene Symbol
TEX33
Entrez Gene ID
767965
Gene Name
testis expressed 33
Gene Aliases
C5H22orf33
Chromosome Location
Chr.5: 75415495 - 75433378 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077033.2 NP_001070501.1 3 - 4 260 86
GenBank mRNA BC109555.1 - 3 - 4 262 86

View Full Product Details