Assay Design



Gene Transcripts

Gene Symbol
CSTF1
Entrez Gene ID
507051
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
-
Chromosome Location
Chr.13: 59540717 - 59554647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077865.2 NP_001071333.1 2 - 3 282 92
GenBank mRNA BC126532.1 - 2 - 3 285 92

View Full Product Details