Assay Design



Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
510275
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
-
Chromosome Location
Chr.19: 44558563 - 44566919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077896.2 NP_001071364.1 1 - 2 105 93
GenBank mRNA BC126834.1 - 1 - 2 106 93

View Full Product Details