Assay Design



Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
510275
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
-
Chromosome Location
Chr.19: 44558563 - 44566919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077896.2 NP_001071364.1 3 - 4 387 117
GenBank mRNA BC126834.1 - 3 - 4 388 117

View Full Product Details