Assay Design



Gene Transcripts

Gene Symbol
KLHL10
Entrez Gene ID
541263
Gene Name
kelch like family member 10
Gene Aliases
-
Chromosome Location
Chr.19: 42024147 - 42037172 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078033.2 NP_001071501.1 3 - 4 1435 63
GenBank mRNA BC126695.1 - 3 - 4 1436 63

View Full Product Details