Assay Design



Gene Transcripts

Gene Symbol
TREM2
Entrez Gene ID
506467
Gene Name
triggering receptor expressed on myeloid cells 2
Gene Aliases
-
Chromosome Location
Chr.23: 15082526 - 15105522 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079580.2 NP_001073048.1 2 - 3 419 114
GenBank mRNA BC122771.1 - 2 - 3 420 114

View Full Product Details