Assay Design



Gene Transcripts

Gene Symbol
CACFD1
Entrez Gene ID
512024
Gene Name
calcium channel flower domain containing 1
Gene Aliases
C11H9orf7
Chromosome Location
Chr.11: 104352228 - 104361245 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080263.1 NP_001073732.1 1 - 2 147 85
GenBank mRNA BC122595.1 - 1 - 2 147 85

View Full Product Details