Assay Design



Gene Transcripts

Gene Symbol
CACFD1
Entrez Gene ID
512024
Gene Name
calcium channel flower domain containing 1
Gene Aliases
C11H9orf7
Chromosome Location
Chr.11: 104352228 - 104361245 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080263.1 NP_001073732.1 3 - 4 348 55
GenBank mRNA BC122595.1 - 3 - 4 348 55

View Full Product Details