Assay Design



Gene Transcripts

Gene Symbol
ITM2C
Entrez Gene ID
539959
Gene Name
integral membrane protein 2C
Gene Aliases
ITM3
Chromosome Location
Chr.2: 118666252 - 118680137 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081538.1 NP_001075007.1 5 - 6 779 80
GenBank mRNA BC133321.1 - 5 - 6 779 80

View Full Product Details