Assay Design



Gene Transcripts

Gene Symbol
C11H2orf49
Entrez Gene ID
540024
Gene Name
chromosome 11 C2orf49 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 9256204 - 9266831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081539.1 NP_001075008.1 2 - 3 319 115
GenBank mRNA BC133333.1 - 2 - 3 319 115

View Full Product Details