Assay Design
Gene Transcripts
- Gene Symbol
- PAGE4
- Entrez Gene ID
-
505725
- Gene Name
- PAGE family member 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 87516566 - 87520757 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001081580.2 |
NP_001075049.1 |
1 - 2 |
104 |
86 |
GenBank mRNA |
BC133479.1 |
- |
1 - 2 |
105 |
86 |
View Full Product Details