Assay Design



Gene Transcripts

Gene Symbol
SGO1
Entrez Gene ID
614056
Gene Name
shugoshin 1
Gene Aliases
-
Chromosome Location
Chr.1: 157392690 - 157411178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081617.1 NP_001075086.1 3 - 4 431 124
GenBank mRNA BC133396.1 - 3 - 4 431 124

View Full Product Details