Assay Design



Gene Transcripts

Gene Symbol
SERPINA3-8
Entrez Gene ID
505820
Gene Name
SERPINA3-8
Gene Aliases
-
Chromosome Location
Chr.21: 59317212 - 59326423 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081712.1 NP_001075181.1 2 - 3 678 120
GenBank mRNA BC149427.1 - 2 - 3 728 120

View Full Product Details