Assay Design



Gene Transcripts

Gene Symbol
CMTR1
Entrez Gene ID
509620
Gene Name
cap methyltransferase 1
Gene Aliases
FTSJD2, MTr1
Chromosome Location
Chr.23: 11318812 - 11372352 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082430.2 NP_001075899.1 5 - 6 652 101
GenBank mRNA BC133559.1 - 5 - 6 653 101

View Full Product Details