Assay Design



Gene Transcripts

Gene Symbol
STX6
Entrez Gene ID
510909
Gene Name
syntaxin 6
Gene Aliases
-
Chromosome Location
Chr.16: 62101080 - 62148223 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082431.1 NP_001075900.1 6 - 7 787 89
GenBank mRNA BC133525.1 - 6 - 7 787 89

View Full Product Details