Assay Design



Gene Transcripts

Gene Symbol
ADPGK
Entrez Gene ID
518158
Gene Name
ADP dependent glucokinase
Gene Aliases
ADP-GK
Chromosome Location
Chr.10: 19545919 - 19578555 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082438.2 NP_001075907.1 3 - 4 581 92
GenBank mRNA BC133568.1 - 3 - 4 582 92

View Full Product Details