Assay Design



Gene Transcripts

Gene Symbol
TSNARE1
Entrez Gene ID
535306
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 1901658 - 2019964 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082446.2 NP_001075915.1 2 - 3 162 79
GenBank mRNA BC133610.1 - 2 - 3 164 79

View Full Product Details