Assay Design



Gene Transcripts

Gene Symbol
CCNI
Entrez Gene ID
504847
Gene Name
cyclin I
Gene Aliases
-
Chromosome Location
Chr.6: 91881487 - 91920418 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083373.1 NP_001076842.1 5 - 6 1045 90
GenBank mRNA BC114754.1 - 5 - 6 1045 90

View Full Product Details