Assay Design



Gene Transcripts

Gene Symbol
TMEM17
Entrez Gene ID
514758
Gene Name
transmembrane protein 17
Gene Aliases
-
Chromosome Location
Chr.11: 61003739 - 61011375 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083433.1 NP_001076902.1 2 - 3 215 123
GenBank mRNA BC123468.1 - 2 - 3 215 123

View Full Product Details