Assay Design



Gene Transcripts

Gene Symbol
APOA5
Entrez Gene ID
538914
Gene Name
apolipoprotein A5
Gene Aliases
-
Chromosome Location
Chr.15: 27443933 - 27446597 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083492.2 NP_001076961.1 1 - 2 70 99
GenBank mRNA BC123517.1 - 1 - 2 71 99

View Full Product Details