Assay Design



Gene Transcripts

Gene Symbol
UBE2M
Entrez Gene ID
613343
Gene Name
ubiquitin conjugating enzyme E2 M
Gene Aliases
-
Chromosome Location
Chr.18: 65779276 - 65782514 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083503.1 NP_001076972.1 4 - 5 861 90
GenBank mRNA BC133501.1 - 4 - 5 861 90

View Full Product Details