Assay Design



Gene Transcripts

Gene Symbol
CERS1
Entrez Gene ID
615296
Gene Name
ceramide synthase 1
Gene Aliases
LASS1
Chromosome Location
Chr.7: 4313437 - 4332887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083514.2 NP_001076983.1 4 - 5 769 104
GenBank mRNA BC114845.1 - 4 - 5 770 104

View Full Product Details