Assay Design



Gene Transcripts

Gene Symbol
TFF2
Entrez Gene ID
616105
Gene Name
trefoil factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 142614739 - 142618005 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083521.1 NP_001076990.1 2 - 3 259 69
GenBank mRNA BC114896.1 - 2 - 3 259 69

View Full Product Details