Assay Design



Gene Transcripts

Gene Symbol
PLEKHB2
Entrez Gene ID
783000
Gene Name
pleckstrin homology domain containing B2
Gene Aliases
-
Chromosome Location
Chr.2: 1920516 - 1969797 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083529.1 NP_001076998.1 6 - 7 472 74
GenBank mRNA BC123674.1 - 6 - 7 472 74

View Full Product Details