Assay Design



Gene Transcripts

Gene Symbol
GALNT9
Entrez Gene ID
504951
Gene Name
polypeptide N-acetylgalactosaminyltransferase 9
Gene Aliases
WBSCR17
Chromosome Location
Chr.17: 44853960 - 44973834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083641.1 NP_001077110.1 6 - 7 1468 123
GenBank mRNA BC134626.1 - 6 - 7 1468 123

View Full Product Details