Assay Design



Gene Transcripts

Gene Symbol
TM9SF1
Entrez Gene ID
539478
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
-
Chromosome Location
Chr.10: 20961901 - 20967166 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083743.1 NP_001077212.1 3 - 4 1098 56
GenBank mRNA BC134551.1 - 3 - 4 1098 56

View Full Product Details