Assay Design



Gene Transcripts

Gene Symbol
SH3BGRL2
Entrez Gene ID
785404
Gene Name
SH3 domain binding glutamate rich protein like 2
Gene Aliases
-
Chromosome Location
Chr.9: 19256542 - 19330621 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083791.1 NP_001077260.1 1 - 2 207 78
GenBank mRNA BC134511.1 - 1 - 2 207 78

View Full Product Details