Assay Design



Gene Transcripts

Gene Symbol
CENPP
Entrez Gene ID
504630
Gene Name
centromere protein P
Gene Aliases
-
Chromosome Location
Chr.8: 83951574 - 84187170 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105615.1 NP_001099085.1 1 - 2 122 109
GenBank mRNA BC134699.1 - 1 - 2 122 109

View Full Product Details