Assay Design



Gene Transcripts

Gene Symbol
PLP1
Entrez Gene ID
281410
Gene Name
proteolipid protein 1
Gene Aliases
PLP
Chromosome Location
Chr.X: 52544092 - 52560716 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174149.4 NP_776574.2 1 - 2 79 64
GenBank mRNA BC103102.1 - 1 - 2 146 64
BC133388.1 - 1 - 2 79 64

View Full Product Details