Assay Design



Gene Transcripts

Gene Symbol
CDK5R1
Entrez Gene ID
282173
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Gene Aliases
P25/P35
Chromosome Location
Chr.19: 17694835 - 17696848 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174512.3 NP_776937.2 1 - 2 54 122
GenBank mRNA BC122779.1 - 1 - 2 123
X78934.1 - - 54 122

View Full Product Details