Assay Design
Gene Transcripts
- Gene Symbol
- SLC2A3
- Entrez Gene ID
-
282358
- Gene Name
- solute carrier family 2 member 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 101438340 - 101451254 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174603.3 |
NP_777028.2 |
3 - 4 |
383 |
69 |
GenBank mRNA |
BC133291.1 |
- |
3 - 4 |
383 |
69 |
View Full Product Details