Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181002.3 | NP_851345.1 | 2 - 3 | 2010 | 71 |
GenBank mRNA | AB019242.1 | - | 2 - 3 | 2081 | 71 |
BC140657.1 | - | 2 - 3 | 2011 | 71 | |
BT021702.1 | - | 2 - 3 | 1954 | 71 | |
Y15774.1 | - | - | 1900 | 71 |