Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181002.3 | NP_851345.1 | 3 - 4 | 2132 | 78 |
GenBank mRNA | AB019242.1 | - | 3 - 4 | 2203 | 78 |
BC140657.1 | - | 3 - 4 | 2133 | 78 | |
BT021702.1 | - | 3 - 4 | 2076 | 78 | |
Y15774.1 | - | - | 2022 | 78 |