Assay Design



Gene Transcripts

Gene Symbol
FAM50A
Entrez Gene ID
515539
Gene Name
family with sequence similarity 50 member A
Gene Aliases
-
Chromosome Location
Chr.X: 37504050 - 37509805 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034496.2 NP_001029668.2 12 - 13 1081 59
GenBank mRNA BC103028.1 - 11 - 12 1358 59
CO887857.1 - 4 - 5 301 59

View Full Product Details