Assay Design



Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
515913
Gene Name
thymus, brain and testes associated
Gene Aliases
C28H10orf27, SPATIAL
Chromosome Location
Chr.28: 26937387 - 26952734 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038121.2 NP_001033210.2 1 - 2 31 73
GenBank mRNA BC110031.1 - 1 - 2 32 73

View Full Product Details