Assay Design



Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
515913
Gene Name
thymus, brain and testes associated
Gene Aliases
C28H10orf27, SPATIAL
Chromosome Location
Chr.28: 26937387 - 26952734 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038121.2 NP_001033210.2 5 - 6 739 63
GenBank mRNA BC110031.1 - 5 - 6 740 63
CK774174.1 - 4 - 5 507 63

View Full Product Details