Assay Design



Gene Transcripts

Gene Symbol
HM13
Entrez Gene ID
512534
Gene Name
histocompatibility minor 13
Gene Aliases
-
Chromosome Location
Chr.13: 61107897 - 61146837 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046124.2 NP_001039589.2 11 - 12 1148 68
GenBank mRNA BC112454.1 - 11 - 12 1130 68

View Full Product Details