Assay Design



Gene Transcripts

Gene Symbol
PAQR6
Entrez Gene ID
516498
Gene Name
progestin and adipoQ receptor family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 14545370 - 14551100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046225.2 NP_001039690.2 2 - 3 200 73
GenBank mRNA BC112862.1 - 2 - 3 143 73
DV879653.1 - 2 - 3 157 73
DV884229.1 - 2 - 3 151 73
EE359306.1 - 2 - 3 200 73

View Full Product Details