Assay Design



Gene Transcripts

Gene Symbol
PAQR6
Entrez Gene ID
516498
Gene Name
progestin and adipoQ receptor family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 14545370 - 14551100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046225.2 NP_001039690.2 5 - 6 667 70
GenBank mRNA DV874964.1 - 2 - 3 159 70
DV879653.1 - 5 - 6 624 70
DV884229.1 - 5 - 6 618 70

View Full Product Details