Assay Design



Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
513572
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
-
Chromosome Location
Chr.7: 96304223 - 96357020 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075628.2 NP_001069096.2 15 - 16 2508 96
GenBank mRNA BC149475.1 - 15 - 16 2508 96

View Full Product Details