Assay Design



Gene Transcripts

Gene Symbol
EPHX4
Entrez Gene ID
524246
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7
Chromosome Location
Chr.3: 51335351 - 51372381 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075855.3 NP_001069323.2 4 - 5 671 114
GenBank mRNA DT720346.1 - 4 - 5 506 114

View Full Product Details