Assay Design



Gene Transcripts

Gene Symbol
LAS1L
Entrez Gene ID
507924
Gene Name
LAS1 like ribosome biogenesis factor
Gene Aliases
-
Chromosome Location
Chr.X: 94949980 - 94970019 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076815.2 NP_001070283.2 7 - 8 1122 63
GenBank mRNA BC149210.1 - 7 - 8 1122 63

View Full Product Details